missing translation for 'onlineSavingsMsg'
Learn More

CD10 Mouse anti-Human, PE-Cy5, Clone: HI10A, BD

Mouse Monoclonal Antibody

Marca:  BD Biosciences 555376

Detalles adicionales : Peso : 0.02000kg

 Ver más versiones de este producto

Código de producto. 15898108

  • 520.00€ / cada uno
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

The HI10a monoclonal antibody specifically binds to CD10 which is also known as Neutral endopeptidase (NEP), Enkephalinase, Atriopeptidase, and Neprilysin. CD10 is encoded by MME (membrane metallo-endopeptidase). CD10 is a 100 kDa type II transmembrane glycoprotein that has neutral endopeptidase activity and is otherwise known as the Common Acute Lymphoblastic Leukemia Antigen (CALLA). CD10 is expressed on a wide variety of normal and neoplastic cell types. Normal cells expressing CD10 include granulocytes, bone marrow stromal cells, a subset of B-cell progenitors, germinal center B cells and fibroblasts. This cell surface metalloendopeptidase inactivates a number of signaling molecules and serves as a major regulator in the nervous, immune and other systems.

Especificaciones

Especificaciones

CD10
Monoclonal
PE-Cyanine5
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
HI10A
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Acute CALLA Leukemia Blast Cells
100 Tests
Immunology
Human
IgG1 κ
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only.