missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLDN1, Human, IP-WB Antibody Pair (PW1), Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Marca: Abnova H00009076-PW1.1Set
Includes: Antibody pair for IP: rabbit polyclonal anti-CLDN1 (300μL), Antibody pair for WB: mouse polyclonal anti-CLDN1 (50μL)
Especificaciones
CLDN1 | |
Monoclonal/Polyclonal | |
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot. | |
CLD1/ILVASC/SEMP1 | |
Mouse, Rabbit | |
RUO | |
9076 | |
Human | |
Liquid |
Immunoprecipitation, Western Blot | |
Unconjugated | |
CLDN1 | |
CLDN1 | |
1 Set | |
Primary | |
Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-CLDN1 (300 ul) 2. Antibody pair for WB: mouse polyclonal anti-CLDN1 (50 ul) | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Sugerencias de productos
Los clientes que vieron este artículo también vieron
Viewing 1-5 of 10
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
CLDN1, Human, IP-WB Antibody Pair (PW1), Abnova™ > 1 Set
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido