missing translation for 'onlineSavingsMsg'
Learn More

anti-IgG2a κ, Rat, APC, Clone: R35-95, Isotype Control, BD

Marca:  BD Biosciences 553932

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15877478

  • 600.00€ / cada uno
Fecha estimada de envío: 12-08-2024
para ver el stock
Añadir a la cesta

Descripción

Descripción

The R35-95 hybridoma was generated by hybridization of Y3 myeloma cells with spleen cells from LOU rats immunized with mouse immunoglobulins. The R35-95 hybridoma produces rat IgG2a, κ immunoglobulin that has no measurable reactivity with mouse immunoglobulins. The R35-95 immunoglobulin was selected as an isotype control following screening for low background binding on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only.

Host Species: Rat
Clone: R35-95
Isotype: IgG2a κ
Immunogen: Mouse Pooled Immunoglobulin

Flow Cytometry, Intracellular Staining, Isotype Control

Especificaciones

Especificaciones

Flow Cytometry
0.2mg/mL
Rat
Affinity Purified
Undiluted at 4°C
R35-95
APC
Mouse Pooled Immunoglobulin
0.1 mg
IgG2a κ
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales