Learn More
IRF3, Human, IP-WB Antibody Pair (PW3), Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Marca:  Abnova H00003661-PW3.1Set
Código de producto. 16102274
  • 809.00€ / 1 set
Explore más ofertas especiales
Includes: Antibody pair for IP: rabbit polyclonal anti-IRF3 (300μL), Antibody pair for WB: mouse purified polyclonal anti-IRF3 (50μg)
Especificaciones
IRF3
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
IRF3
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-IRF3 (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-IRF3 (50 ug)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Immunoprecipitation, Western Blot
Unconjugated
IRF3
Mouse, Rabbit
RUO
3661
Human
Liquid
Documentos
One moment while we fetch your results.
Certificados
Ofertas especiales
Ofertas especiales
Título del producto
IRF3, Human, IP-WB Antibody Pair (PW3), Abnova™ > 1 Set
For Research Use Only