Learn More
IRF3, Human, IP-WB Antibody Pair (PW3), Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Marca: Abnova H00003661-PW3.1Set
Includes: Antibody pair for IP: rabbit polyclonal anti-IRF3 (300μL), Antibody pair for WB: mouse purified polyclonal anti-IRF3 (50μg)
Especificaciones
IRF3 | |
Monoclonal/Polyclonal | |
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot. | |
IRF3 | |
1 Set | |
Primary | |
Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-IRF3 (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-IRF3 (50 ug) | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Immunoprecipitation, Western Blot | |
Unconjugated | |
IRF3 | |
Mouse, Rabbit | |
RUO | |
3661 | |
Human | |
Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
IRF3, Human, IP-WB Antibody Pair (PW3), Abnova™ > 1 Set