Learn More
Banoxantrone dihydrochloride
Prodrug topoisomerase II inhibitor
Marca:  Tocris Bioscience 4219/5
 Ver más versiones de este producto
Código de producto. 18706416
  • 127.00€ / 5 miligramos
Explore más ofertas especiales
Descripción
Descripción
Banoxantrone dihydrochloride is a bioreductive prodrug; activated by hypoxia to yield the topoisomerase II inhibitor AQ4. Displays little to no cytotoxic activity against 60 tumor cell lines tested under oxic conditions (IC50 >100 μM). Exhibits anti-tumor efficacy in vivo when combined with oxic cell cytotoxins; enhances the anti-tumor effect of cyclophosphamide (Cat. No. 4091) in a mouse tumor model.
Especificaciones
Especificaciones
1,4-Bis[[2-(dimethyloxidoamino)ethyl]amino]-5,8-dihydroxy-9,10-anthracenedione dihydrochloride
>98%
C22H28N4O6.2HCl
252979-56-9
5 mg
Observed MW: 517.4