Learn More
Banoxantrone dihydrochloride
Prodrug topoisomerase II inhibitor
Brand: Tocris Bioscience 4219/5
This item is not returnable.
View return policy
Description
Banoxantrone dihydrochloride is a bioreductive prodrug; activated by hypoxia to yield the topoisomerase II inhibitor AQ4. Displays little to no cytotoxic activity against 60 tumor cell lines tested under oxic conditions (IC50 >100 μM). Exhibits anti-tumor efficacy in vivo when combined with oxic cell cytotoxins; enhances the anti-tumor effect of cyclophosphamide (Cat. No. 4091) in a mouse tumor model.Specifications
1,4-Bis[[2-(dimethyloxidoamino)ethyl]amino]-5,8-dihydroxy-9,10-anthracenedione dihydrochloride | |
>98% | |
C22H28N4O6.2HCl |
252979-56-9 | |
5 mg | |
Observed MW: 517.4 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction