Learn More
BD Difco™ Microbial Content Test Agar
Description
Certifications
CCAM
Specifications
Specifications
| Certifications/Compliance | CCAM |
| Content And Storage | 2°C to 8°C |
| Form | Dehydrated |
| Media Type | Microbial Content Test Agar |
| For Use With (Application) | Detection and enumeration of microorganisms |
| Packaging Type | Bottle |
| Quantity | 500 g |