Learn More
ACROBiosystems Biotinylated Human 4-1BB Ligand / TNFSF9 Protein, Fc, Avitag™
Biotinylated Protein
Marca: ACROBiosystems 41L-H82F9-25UG
Detalles adicionales : Peso : 0.00001kg
Descripción
Tumor necrosis factor ligand superfamily member 9 (4-1BBL) is also known as 4-1BB ligand, CD137L or TNFSF9, which is a cytokine that binds to TNFRSF9. 4-1BBL is the high affinity ligand of 4-1BB. 4-1BBL induces the proliferation of activated peripheral blood T-cells. Also, 4-1BBL may have a role in activation-induced cell death (AICD). Furthermore, 4-1BBL may play a role in cognate interactions between T-cells and B-cells/macrophages. As for diseases, 4-1BBL is involved in cancers, infectious diseases and autoimmune diseases.
This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 50.2 kDa. The protein migrates as 50-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
NP_003802.1 | |
25 μg | |
Ala 50 - Glu 254 | |
4-1 BB Ligand,TNFSF9,CD137L | |
Precision Avi | |
Fc Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
50.2 kDa | |
Biotinylated Human 4-1 BB Ligand, Fc,Avitag (41 L-H82F9) is expressed from human 293 cells (HEK293). It contains AA Ala 50 - Glu 254 (Accession # P41273-1). | |
-20°C | |
4-1 BB Ligand | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |