missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Biotinylated Human 4-1BB Ligand / TNFSF9 Protein, Fc, Avitag™
Biotinylated Protein
423.38€ - 1862.87€
Especificaciones
Número de acceso | NP_003802.1 |
---|---|
Peso molecular | 50.2 kDa |
Fuente | Biotinylated Human 4-1 BB Ligand, Fc,Avitag (41 L-H82F9) is expressed from human 293 cells (HEK293). It contains AA Ala 50 - Glu 254 (Accession # P41273-1). |
Inmunógeno | Ala 50 - Glu 254 |
Requisitos de almacenamiento | -20°C |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15885867
|
ACROBiosystems
41L-H82F9-200UG |
200 μg |
1862.87€
200 microgramos |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
15895867
|
ACROBiosystems
41L-H82F9-25UG |
25 μg |
423.38€
25 microgramos |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
Tumor necrosis factor ligand superfamily member 9 (4-1BBL) is also known as 4-1BB ligand, CD137L or TNFSF9, which is a cytokine that binds to TNFRSF9. 4-1BBL is the high affinity ligand of 4-1BB. 4-1BBL induces the proliferation of activated peripheral blood T-cells. Also, 4-1BBL may have a role in activation-induced cell death (AICD). Furthermore, 4-1BBL may play a role in cognate interactions between T-cells and B-cells/macrophages. As for diseases, 4-1BBL is involved in cancers, infectious diseases and autoimmune diseases.
This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 50.2 kDa. The protein migrates as 50-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
NP_003802.1 | |
Biotinylated Human 4-1 BB Ligand, Fc,Avitag (41 L-H82F9) is expressed from human 293 cells (HEK293). It contains AA Ala 50 - Glu 254 (Accession # P41273-1). | |
-20°C | |
4-1 BB Ligand | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
50.2 kDa | |
Ala 50 - Glu 254 | |
4-1 BB Ligand,TNFSF9,CD137L | |
Precision Avi | |
Fc Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |