missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD13 Mouse anti-Human, Clone: 2D8D11, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66211-1-IG-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
CD13 (ANPEP) is a 150-170 kDa type II transmembrane zinc-binding ectopeptidase expressed on various cell types. CD13 is a metalloprotease that preferentially catalyzes removal of neutral amino acids from small peptides, thus activating or inactivating bioactive peptides. CD13 also has a role in extracellular matrix degradation, antigen processing and signal transduction, is important in inflammatory responses, regulates intercellular contact, cell motility and vascularization. CD13 is expressed by granulocytes, monocytes, myeloid leukemia cells, and is involved in protection of leukemic cells against apoptosis and its expression associated with poor prognosis of carcinomas. CD13 functions as an aminopeptidase enzyme and is also alpha receptor for coronavirus.Especificaciones
CD13 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P15144 | |
ANPEP | |
CD13 Fusion Protein Ag5976 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
2D8D11 | |
Unconjugated | |
ANPEP | |
Alanyl aminopeptidase, Aminopeptidase M, Aminopeptidase N, ANPEP, AP M, AP N, APN, CD13, gp150, hAPN, LAP1, Microsomal aminopeptidase, p150, PEPN | |
Mouse | |
Protein A | |
RUO | |
290 | |
-20°C | |
Liquid |