missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD366 (TIM3) Monoclonal Antibody (F38-2E2), Brilliant Violet™ 605, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
175.00€ - 358.00€
Especificaciones
Antígeno | CD366 (TIM3) |
---|---|
Clon | F38-2E2 |
Concentración | 5 μL/Test |
Aplicaciones | Flow Cytometry |
Clasificación | Monoclonal |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
17914332
|
Invitrogen
406-3109-42 |
100 Tests |
358.00€
paquete de 100 |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
17934222
|
Invitrogen
406-3109-41 |
25 Tests |
175.00€
paquete de 25 |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Descripción
TIM3 (Hepatitis A virus cellular receptor 2, HAVCR2, T-cell immunoglobulin, mucin-dmain containing-3) is a 281 amino acid long, Type-1 Th1- specific cell surface glycoprotein expressed on terminally differentiated CD4+Th1 and CD8+Tc1 cells. TIM3 consists of an IgV-like domain, a mucin-like domain in the extracellular region, and a conserved Tyrosine phosphorylation motif in the cytoplasmic region. TIM3 is involved in macrophage activation and induction of autoimmune diseases. Further, TIM3 down-regulates aggressive Th1-mediated immune responses and facilitates in the development of immune tolerance. Pathological significance of TIM3 has been attributed to Experimental autoimmune encephalomyelitis (EAE), a Th-1 dependent autoimmune disease, and also enhances the severity of experimental autoimmune encephalomyelitis in mice.Especificaciones
CD366 (TIM3) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide, pH 7.2 | |
CD366, FLJ14428, Havcr2, HAVcr-2, Hepatitis A virus cellular receptor 2, hepatitis A virus cellular receptor 2 homolog, kidney injury molecule-3, KIM-3, sCD366, soluble CD366, soluble TIM 3, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain containing 3, T-cell immunoglobulin and mucin domain-containing protein 3, T-cell immunoglobulin mucin family member 3, T-cell immunoglobulin mucin receptor 3, T-cell membrane protein 3, Tim3, TIM-3, TIMD3, TIMD-3 | |
Havcr2 | |
Primary | |
4°, C, store in dark, DO NOT FREEZE! |
F38-2E2 | |
Flow Cytometry | |
Brilliant Violet 605 | |
Mouse | |
Human | |
Q8TDQ0 | |
84868 | |
IgG1 κ | |
Affinity chromatography | |
Havcr2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title