missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45RO Mouse anti-Human, BB515, Clone: UCHL1, BD Horizon™
Mouse Monoclonal Antibody
Marca: BD Biosciences 564530
Detalles adicionales : Peso : 0.01000kg
Descripción
CD45RO Monoclonal antibody specifically detects CD45RO in Human samples. It is validated for Flow cytometry.
CD45RO Monoclonal antibody specifically detects CD45RO in Human samples. It is validated for Flow cytometry.Especificaciones
CD45RO | |
Monoclonal | |
Brilliant Blue 515 (BB515) | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
25 Tests | |
Cell Biology | |
Human | |
Purified |
Flow Cytometry | |
UCHL1 | |
Vol. Per test: 5μL | |
CD45R; PTPRC; LCA; Leukocyte common antigen; GP180; LY5; T200 | |
Human IL-2-dependent T-cell line | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed. | |
IgG2a κ |
For Research Use Only.