missing translation for 'onlineSavingsMsg'
Learn More

CD45RO Mouse anti-Human, BB515, Clone: UCHL1, BD Horizon™

Mouse Monoclonal Antibody

Marca:  BD Biosciences 564529

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15893699

  • 447.00€ / cada uno
Fecha estimada de envío: 12-08-2024
para ver el stock

Descripción

Descripción

CD45RO Monoclonal antibody specifically detects CD45RO in Human samples. It is validated for Flow cytometry.

CD45RO Monoclonal antibody specifically detects CD45RO in Human samples. It is validated for Flow cytometry.
Especificaciones

Especificaciones

CD45RO
Monoclonal
Brilliant Blue 515 (BB515)
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse
100 Tests
Cell Biology
Human
Purified
Flow Cytometry
UCHL1
Vol. Per test: 5μL
CD45R; PTPRC; LCA; Leukocyte common antigen; GP180; LY5; T200
Human IL-2-dependent T-cell line
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed.
IgG2a κ
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only.