Learn More
CD66b Mouse anti-Human, FITC, Clone: G10F5, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 561927
Detalles adicionales : Peso : 0.02000kg
Descripción
The G10F5 monoclonal antibody specifically binds to CD66b, also known as Carcinoembryonic antigen-related cell adhesion molecule 8 (CEACAM8). CD66b is a glycosylphosphatidylinositol (GPI) linked protein with amolecular weight of 100 kDa expressed on granulocytes. This molecule was previously clustered as CD67 in the Fourth Human Leucocyte DifferentiationAntigen (HLDA) Workshop and renamed CD66b in the Fifth HLDA Workshop. CD66b is a member of the carcinoembryonic antigen (CEA)-like glycoprotein family present on granulocytes and referred to as non-specific crossreacting antigens (NCA). Granulocyte activation induced with soluble stimulators (calcium ionophore, phorbol myristate acetate, N-formylmethionyl- leucyl-phenylalanine) results in release and increased expression of NCA. Findings suggest that these molecules may play a role in phagocytosis, chemotaxis and adherence.
Especificaciones
CD66b | |
Monoclonal | |
FITC | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgM κ |
Flow Cytometry | |
G10F5 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human Granulocytes | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.