Learn More
Eurofins DiscoverX CDK4/Cyclin D3 Protein, Active
N-terminal GST-tagged, recombinant, human CDK4 full length and N-terminal GST-tagged, recombinant, human cyclin D3 full length co-expressed by baculovirus in Sf21 insect cells. for use in Kinase Assays
Marca: Eurofins DiscoverX 14-957M
Descripción
CDK4 (Cyclin-dependent kinase 4) forms a complex with type D cyclins such as D1, D2 and D3 and when the cellular levels of cyclin D peak there is a shortening of the G1 phase and rapid entry into S phase of cell cycle progression. The substrate for cdk4/cyclin D is Rb, and phosphorylation of Rb results in dissociation of the E2F complex from Rb. The free E2F proteins in turn stimulate transcription of genes required for DNA synthesis. Cellular inhibition of CDK4/cyclin D3 activity is achieved by its inhibitory proteins, i.e. p16, p21 and p27. Inhibition of CDK4/CCND3 may be used in treatment of uncontrolled cell proliferation, tumor heterogeneity, invasion and metastasis.
GenBank NM_000075.3 / GenBank M90814.1
Especificaciones
mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% β-mercaptoethanol. Frozen solution. | |
250 μg | |
Recombinant | |
Recombinant enzyme expressed in Sf21 insect cells | |
Combined ≥72% ratio 2-3:1 CDK4:cyclinD3 |
61 kDa (CDK4) and 59 kDa (Cyclin D3) | |
Human | |
N-terminal GST / N-terminal GST | |
Full-length | |
CDK4/Cyclin D3, active |