Learn More
Corning™ Fetal Bovine Serum, Regular (Heat Inactivated)
BIOPHARMAGREENER_CHOICE
FBS is a commonly used cell culture media supplement which helps sustain cell growth and satisfy metabolic requirements.
Marca:  Corning™ 35-011-CV
Código de producto. 15353681
  • 660.00€ / 500 ml
Explore más ofertas especiales
Descripción
Descripción
  • Sera is aseptically collected and processed in a cGMP facility
  • Each batch of raw serum is screened for endotoxin, residual hemoglobin concentration, and microbiological quality before being used in production
  • Batches of raw serum are pooled and continuously mixed prior to filtration to ensure consistent quality throughout the entire lot
  • Final lots are tested for endotoxin, hemoglobin, pH, mycoplasma, and growth promotion. Biochemical assays and virus testing are also available upon request
  • Regular FBS is sourced from herds in USDA approved countries (ex: Mexico, Honduras, Costa Rica, Nicaragua, etc.)
Certificados
Se requiere un número de lote para mostrar resultados para certificados. Para encontrar su número de lote en pedidos anteriores, utilice el área de estado de pedidos.
3 Resultados encontrados
Número de lote Tipo de certificado Fecha Catalog Number
Número de lote03724002 Tipo de certificadoCertificado de calidad Fecha19/03/2025 Catalog Number35-011-CV
Número de lote23623001 Tipo de certificadoCertificado de calidad Fecha06/08/2024 Catalog Number35-011-CV
Número de lote13823001 Tipo de certificadoCertificado de calidad Fecha03/07/2024 Catalog Number35-011-CV
Ofertas especiales
Ofertas especiales
Título del producto
Corning™ Fetal Bovine Serum, Regular (Heat Inactivated) > Heat Inactivated; USDA Approved; 500 mL