missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CSNK1A1L, GST Tag Recombinant Protein
This CSNK1A1L Recombinant Human Protein (full length) was expressed in insect cells.
Marca: Invitrogen™ A32865
Detalles adicionales : Peso : 0.00900kg
Descripción
CSNK1A1L serine/threonine kinase or CK1-alpha dynamically associates with the CBM complex upon T cell receptor engagement to participate in cytokine production and lymphocyte proliferation which has a contrasting role by subsequently promoting the phosphorylation and inactivation of CARMA1. It governs antigen-receptor-induced NF-kappa-B activation and human lymphoma cell survival.- Validated for use with the Z’-LYTE™ assay
- Available in multiple pack sizes for convenient use
Especificaciones
AAH28723 | |
65.4 kDa | |
CK1 Kinase Family | |
Human | |
GST Tag | |
Dry Ice | |
Full Length, Recombinant, Active |
Kinase Assay | |
100 μg | |
Full Length | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol. | |
Insect | |
Liquid |