Learn More
Sartorius Cubis™ II High-Capacity Micro Balance, 36S
The Cubis™ II range of high-capacity micro balances with a maximum load between 32 g and 111 g and a readability between 0.001 mg and 0.002 mg provide the ideal choice for a broad range of applications.
Marca: Sartorius MCA36S-3S00-D HWL
32090.00 EUR hasta el 2025-12-31
Use el código promocional "25339" para aplicar la oferta.
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- Ultra-high resolution with up to 61 million weighing steps (digits).
- End-to-end data integrity.
- 21 CFR Part 11 compliance, integrated audit-trail, state-of-the-art user management.
- Cleanability, part of compliance.
- Error-free operation.
- Individual QApp workflows, motorized auto-leveling.
- Allow upgrades for hardware features like automated draft shields or built-in ionizer.
- Optional inner draft shield for best weighing performance.
Especificaciones
MCA | |
Color Touchscreen | |
36S Weighing Module | |
3.5 sec. | |
9.4 in. | |
11.9 in. |
32 g | |
0.001 mg | |
MCA36S-3S00-D HWL | |
Balance | |
240 mm | |
302 mm |
CE_17937410_EN.pdf
Tamaño : 117.9 kB
Título del producto
Sartorius Cubis™ II High-Capacity Micro Balance, 36S > Cubis™ II High-Capacity Micro Balance; Model: MCA36S-3S00-D HWL
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido