Learn More
Tocris Bioscience™ DCEBIO
Activates Cl- conductance and hKCa3.1 channels
Marca: Tocris Bioscience™ 1422/10
Detalles adicionales : Peso : 0.00970kg
Descripción
DCEBIO is a stimulates Cl- secretion via the activation of hKCa3.1 (IK1) potassium channels and an apical Cl- conductance carried by the CFTR channel. More potent than its analog 1-EBIO (Cat. No. 1041).Especificaciones
5,6-Dichloro-1-ethyl-1,3-dihydro-2H-benzimidazol-2-one | |
>99% | |
Soluble to 100mM in DMSO | |
Observed MW: 231.08 |
60563-36-2 | |
10 mg | |
C9H8Cl2N2O |
For Research Use Only