missing translation for 'onlineSavingsMsg'
Learn More

Dimetilsulfuro, +99 %, Thermo Scientific Chemicals

40.70€ - 101.51€

Identificadores de productos químicos

CAS 75-18-3
Fórmula molecular C2H6S
Peso molecular (g/mol) 62.13
Número MDL MFCD00008562
Clave InChI QMMFVYPAHWMCMS-UHFFFAOYSA-N
Sinónimo dimethyl sulfide, methyl sulfide, methane, thiobis, dimethylsulfide, dimethylsulphide, methyl thioether, 2-thiapropane, dimethyl sulphide, dimethyl thioether, methylthiomethane
PubChem CID 1068
ChEBI CHEBI:17437
SMILES CSC
Ver más especificaciones

Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
11398217
SDS Ver documentos
Thermo Scientific Alfa Aesar
022949.AK
250 mL
40.70€
250 ml
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
11308227
SDS Ver documentos
Thermo Scientific Alfa Aesar
022949.K2
1 L
101.51€
1 litro
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Descripción

Descripción

Dimethyl sulfide is used as a displaceable ligand in chloro(dimethyl sulfide)gold(I) and other coordination compounds, as a reducing agent in ozonolysis reactions as well as a food flavoring component. Further, it is utilized for the preparation of dimethyl sulfoxide, which is a common and important industrial solvent. It finds application in the petroleum industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Especificaciones
Mostrar más
Videos
Seguridad y manipulación
Mostrar más
SDS
Documentos

Documentos

Product Certifications

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto
Dimetilsulfuro, +99 %, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.