Learn More
Nabertherm™ Drying oven
With their maximum working temperature of up to 300 °C and air circulation, the ovens achieve a perfect temperature uniformity which is much better than in ovens of most competitors. They can be used for various applications such as e.g. drying, sterilizing or warm storing.
Marca: Nabertherm™ TR-242BN
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- TMAX: 300 °C
Especificaciones
B410 | |
300 °C | |
2 | |
1000 × 780 × 970 mm |
240 l | |
8 | |
750 × 550 × 600 mm |