missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nabertherm™ Drying oven
With their maximum working temperature of up to 300 °C and air circulation, the ovens achieve a perfect temperature uniformity which is much better than in ovens of most competitors. They can be used for various applications such as e.g. drying, sterilizing or warm storing.
2539.80€ - 9792.00€
Código de producto | Marca | Capacidad (métrico) | Tipo de controlador | Dimensiones (Largo x Ancho x Alto) | Precio | Cantidad y disponibilidad | |||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Capacidad (métrico) | Tipo de controlador | Dimensiones (Largo x Ancho x Alto) | Precio | Cantidad y disponibilidad | |||
15511122
|
Nabertherm™
TR-062BN |
60 l | B410 | 700 × 610 × 710 mm | |||||
15531122
|
Nabertherm™
TR-122BN |
120 l | B410 | 900 × 610 × 860 mm | |||||
15551122
|
Nabertherm™
TR-242BN |
240 l | B410 | 1000 × 780 × 970 mm | |||||
15561122
|
Nabertherm™
TR-242RN |
240 l | R7 | 1000 × 780 × 970 mm | |||||
15571122
|
Nabertherm™
TR-452BN |
450 l | B410 | 1000 × 780 × 1470 mm | |||||
15581122
|
Nabertherm™
TR-452RN |
450 l | R7 | 1000 × 780 × 1470 mm | |||||
15531460
|
Nabertherm™
TR-903BN |
1050 l | B410 | 1470 × 940 × 1920 mm | |||||
Descripción
- Tmax: 300°C
Especificaciones
300 °C |