missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-isocyanato-3-phenylpropionate, 98%, Thermo Scientific™

Product Code. 11333126
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
Product Code. Quantity unitSize
11333126 10 g 10g
11323126 2 g 2g
2 options
This item is not returnable. View return policy

Product Code. 11333126

Brand: Thermo Scientific Alfa Aesar L10993.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87543-80-4
Molecular Formula C12H13NO3
Molecular Weight (g/mol) 219.24
MDL Number MFCD00082663
InChI Key LHLIGPDDSKJRQN-UHFFFAOYNA-N
Synonym ethyl 2-isocyanato-3-phenylpropionate, 2-isocyanato-3-phenylpropionic acid ethyl ester, 2-isocyanato-3-phenyl-propionic acid ethyl ester, ethyl 2-isocyanato-3-phenyl-propanoate, ethyl n-oxomethylidene phenylalaninate, 2-isocyanato-3-phenylpropanoic acid ethyl ester, benzenepropanoic acid, a-isocyanato-, ethyl ester
PubChem CID 12370705
IUPAC Name ethyl 2-isocyanato-3-phenylpropanoate
SMILES CCOC(=O)C(CC1=CC=CC=C1)N=C=O

Specifications

Boiling Point 152°C (10 mmHg)
Refractive Index 1.504
Quantity 10 g
UN Number UN2206
Beilstein 1247955
Sensitivity Moisture sensitive
Formula Weight 219.24
Percent Purity 98%
Chemical Name or Material Ethyl 2-isocyanato-3-phenylpropionate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.