Learn More
Julabo™ TopTech Series Refrigerated/Heating Circulating Baths with ME Circulators
For working temperatures from -50deg.C to +200°C. Julabo™ TopTech Series Refrigerated/Heating Circulating Baths with ME Circulators has connection for external Pt100 sensor for direct measuring and control in external application.
Marca: Julabo™ 9162634
Descripción
- VFD Comfort Display
- PID3 temperature control, stability ±0.01°C
- Keypad for setpoints, warning/safety values and menu functions
- Integrated programmer (1 x 10 steps) with real-time clock
- ATC3 3-Point-Calibration
- Pt100 External sensor connection for measurement and control
- SMART PUMP, electronically adjustable pump stages
- Adjustable high temperature cut-out, visible via display
- Early warning system for low liquid level (DBGM 203 06 059.8)
- RS232 interface for online communication
- Integrated programmer for 10 program steps
- Active Cooling Control
Especificaciones
PID, Cascade | |
Clasificación III (FL) según DIN 12876-1 | |
11 to 16 L/min. | |
VFD | |
2 barbed fittings suitable for tubing 8 and 10mm inner dia. (pump connections M10x1 female) | |
F34-ME | |
58 x 38 x 62 cm | |
23.8 cm | |
14.9 cm | |
42 kg | |
115 V | |
60 Hz |
Interior (L x An x F): 30 x 24 x 15 cm | |
0.45kW at 20°C, 0.32kW at 0°C, 0.14kW at -20°C | |
De 5 °C a 40 °C | |
1 kW | |
0.23 to 0.45 bar | |
±0.01°C | |
38.1 cm | |
61.9 cm | |
41.7 kg | |
115 V 60 Hz 14 A | |
14 A |