Learn More
GAPDH Antibody (GA1R), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Marca:  Novus Biologicals NBP2-37828AF750
Código de producto. 18712783
  • 588.00€ / 0.10 ml
Explore más ofertas especiales
Descripción
Descripción
GAPDH Monoclonal antibody specifically detects GAPDH in Human, Mouse, Rat, Porcine, Bacteria, Chicken, Hamster, Insect, Rabbit, Yeast samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Human, Mouse, Pig, Chicken, Hamster, Rabbit
Certificados
Ofertas especiales
Ofertas especiales
Título del producto
GAPDH Antibody (GA1R), Alexa Fluor™ 750, Novus Biologicals™ > 0.1 mL; Alexa Fluor 750