Learn More
Greiner Bio-One Tubos de suero: Activador de coágulo con separador de gel
Para pruebas de química clínica rutinaria, inmunohematología, serología o inmunología
Marca: Greiner Bio-One 455071
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- Coated with micronized silica particles which activate clotting when tubes are gently inverted
- Tubes with gel contain a barrier gel on the tube bottom that during centrifugation moves upward to the serum-clot interface to form a stable barrier, separating serum from cells
- Clear as glass with equal or better performance
- Molded to be virtually unbreakable, providing a better level of safety than glass
- Tubes have an 18-month shelf life from the date of manufacture
- Pediatric tubes come with white rings for quick identification
- Safety Twist Caps can be removed and replaced several times
- Nonscrew caps can be removed and replaced as often as required
Compliance
Los tubos con separador de gel están aprobados por la FDA 510(k) para el control terapéutico de fármacos (TDM)
Especificaciones
Microscopic Silica Particles with Separation Gel | |
Non-ridged Pull Cap | |
Red with Yellow Ring |
Plastic | |
8 mL |