Learn More
Tocris Bioscience™ GSK 6853
Potent and selective BRPF1 inhibitor
Marca: Tocris Bioscience™ 6198/5
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
GSK 6853 is a potent and selective BRPF1 inhibitor. Exhibits >1600-fold selectivity for BRPF1 over other bromodomains (pIC50 values by TR-FRET are 8.1, 5.1/4.8 and 4.7/<4.3 for BRPF1, BRPF2/3 and BRD4 BD1/2, respectively).Negative Control also available.
Especificaciones
N-[2,3-dihydro-1,3-dimethyl-6-[(2R)-2-methyl-1-piperazinyl]-2-oxo-1H-benzimidazol-5-yl]-2-methoxybenzamide | |
5 mg | |
409.48 |
1910124-24-1 | |
C22H27N5O3 | |
98% |