missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Calcitonin R Antibody, R&D Systems™
Mouse Monoclonal Antibody
Marca: R&D Systems MAB46141-100
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Calcitonin R Monoclonal specifically detects Calcitonin R in Human samples. It is validated for Western Blot, Immunohistochemistry.Especificaciones
Calcitonin R | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
calcitonin receptor, CalcitoninR, CALCR, CRT, CTR, CT-R, CTR1 | |
HEK293 human embryonic kidney cell line transfected with human Calcitonin R, Ala25-Ala474, Accession # NP_001733 | |
100 μg | |
Primary | |
Detects human Calcitonin R in direct ELISAs. | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
462815 | |
Western Blot 2 ug/mL, Immunohistochemistry 5-25 ug/mL | |
NP_001733 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
799 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
IgG2a |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido