missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GTF2I Full-length ORF (AAH04472, 1 a.a. - 274 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00002969-P02.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16165911

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 04-09-2024
para ver el stock

Descripción

Descripción

This gene encodes a multifunctional phosphoprotein with roles in transcription and signal transduction. It is deleted in Williams-Beuren syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at chromosome 7q11.23. Alternative splicing results in multiple transcript variants. Related pseudogenes have been identified on chromosomes 7, 13 and 21. [provided by RefSeq]

Sequence: MAQVAMSTLPVEDEESSESRMVVTFLMSALESMCKELAKSKAEVACIAVYETDVFVVGTERGRAFVNTRKDFQKDFVKYCVEEEEKAAEMHKMKSTTQANRMSVDAVEIETLRKTVEDYFCFCYGKALGKSTVVPVPYEKMLRDQSAVVVQGLPEGVAFKHPENYDLATLKWILENKAGVSFIIKRPFLEPKKHVGGRVMVTDADRSILSPGGSCGPIKVKTEPTEDSGISLEMAAVTVKEESEDPDYYQYNIQGSHHSSEGNEGTEMEVPAEG
Especificaciones

Especificaciones

AAH04472
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
55.88kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BAP-135/BAP135/BTKAP1/DIWS/FLJ38776/FLJ56355/IB291/SPIN/TFII-I/WBS/WBSCR6
GTF2I
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
2969
GTF2I (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue
MAQVAMSTLPVEDEESSESRMVVTFLMSALESMCKELAKSKAEVACIAVYETDVFVVGTERGRAFVNTRKDFQKDFVKYCVEEEEKAAEMHKMKSTTQANRMSVDAVEIETLRKTVEDYFCFCYGKALGKSTVVPVPYEKMLRDQSAVVVQGLPEGVAFKHPENYDLATLKWILENKAGVSFIIKRPFLEPKKHVGGRVMVTDADRSILSPGGSCGPIKVKTEPTEDSGISLEMAAVTVKEESEDPDYYQYNIQGSHHSSEGNEGTEMEVPAEG
RUO
GTF2I
Wheat Germ (in vitro)
GST
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales