Learn More
R&D Systems™ Human M-CSF Quantikine ELISA Kit
Human M-CSF Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Marca: R&D Systems™ SMC00B
Descripción
The Human M-CSF Quantikine ELISA Kit quantifies human M-CSF.
Especificaciones
78.10 to 5000 pg/mL | |
HRP | |
Solid Phase Sandwich ELISA | |
Natural and Recombinant Human M-CSF | |
Human | |
1435 | |
Cell Culture Supernates (100 μL), Serum (100 μL), EDTA Plasma (100 μL), Heparin Plasma (100 μL), Saliva (100 μL), Urine (100 μL) | |
colony stimulating factor 1 (macrophage), CSF1, CSF-1, Lanimostim, macrophage colony stimulating factor, macrophage colony-stimulating factor 1, MCSF, M-CSF, MCSFlanimostim, MGC31930 |
47.3 pg/mL | |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules | |
Cell Culture Supernatant, Plasma (EDTA, Heparin), Saliva, Serum, Urine | |
6 x 96-well Plates | |
ELISA | |
6 Plates | |
Store the unopened product at 2° to 8°C. Do not use past expiration date. |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.