Learn More
Human/Mouse Cleaved Caspase-3 Alexa Fluor 488-conjugated Antibody, R&D Systems™
Rabbit Monoclonal Antibody has been used in 2 publications
Marca: R&D Systems IC835G-100
Descripción
Caspase-3 Monoclonal specifically detects Caspase-3 in Human, Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.
Especificaciones
Caspase-3 | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
Apopain, apoptosis-related cysteine protease, CASP3, CASP-3, caspase 3, apoptosis-related cysteine peptidase, caspase-3, Caspase3, CPP32, CPP-32, CPP32B, CPP32SREBP cleavage activity 1, Cysteine protease CPP32, EC 3.4.22, EC 3.4.22.56, LICE-1, PARP cleavage protease, procaspase3, Protein Yama, SCA-1, Yama | |
Rabbit | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
836 | |
Mouse, Human | |
Supernatant |
Flow Cytometry | |
269518 | |
Intracellular Staining by Flow Cytometry 5 uL/10^6 cells | |
U26943 | |
CASP3 | |
KLH-conjugated human Caspase-3 synthetic peptide CRGTELDCGIETD Accession No. U26943 | |
100 Tests | |
Primary | |
Detects human and mouse Caspase-3 cleaved at Asp175. No cross-reactivity was detected with the full-length procaspase-3 or other caspases. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.