missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PAPSS2 Partial ORF (NP_004661, 513 a.a. - 612 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00009060-Q01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16169835

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 04-09-2024
para ver el stock

Descripción

Descripción

Sulfation is a common modification of endogenous (lipids, proteins, and carbohydrates) and exogenous (xenobiotics and drugs) compounds. In mammals, the sulfate source is 3'-phosphoadenosine 5'-phosphosulfate (PAPS), created from ATP and inorganic sulfate. Two different tissue isoforms encoded by different genes synthesize PAPS. This gene encodes one of the two PAPS synthetases. Defects in this gene cause the Pakistani type of spondyloepimetaphyseal dysplasia. Two alternatively spliced transcript variants that encode different isoforms have been described for this gene. [provided by RefSeq]

Sequence: DPAGMPHPETKKDLYEPTHGGKVLSMAPGLTSVEIIPFRVAAYNKAKKAMDFYDPARHNEFDFISGTRMRKLAREGENPPDGFMAPKAWKVLTDYYRSLE
Especificaciones

Especificaciones

NP_004661
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
DPAGMPHPETKKDLYEPTHGGKVLSMAPGLTSVEIIPFRVAAYNKAKKAMDFYDPARHNEFDFISGTRMRKLAREGENPPDGFMAPKAWKVLTDYYRSLE
RUO
PAPSS2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
9060
PAPSS2 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ATPSK2/SK2
PAPSS2
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales