Learn More
Gibco™ Human Relaxin-2 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Marca: Gibco™ 130-15-500UG
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Relaxin 1 (also referred to as Relaxin or RLXH1) is a peptide hormone produced by the corpora lutea of the ovary during pregnancy in many mammalian species, including human. The secretion of the hormone into the blood stream just before parturition results in a marked softening and lengthening of the pubic symphysis and a softening of the cervix, which facilitates the birth process. By inhibiting uterine contractions, Relaxin 1 may influence the timing of parturition. Like insulin, Relaxin 1 consists of two peptide chains, A and B, covalently linked by disulfide bonds. By further analogy to insulin, the two peptides are synthesized as a single-chain precursor polypeptide with the B chain at the amino-terminus. The gene that encodes the human Relaxin 1 protein maps to chromosome 9. Relaxin 2, a related protein, is selectively expressed in the ovary during pregnancy. The gene that encodes the human Relaxin 2 protein also maps to chromosome 9.Especificaciones
P04090 | |
protein with no preservative | |
6 | |
500 μg | |
-20°C | |
<1 EU/μg | |
RLN2 | |
Unconjugated | |
A-Chain: QLYSALANKC CHVGCTKRSL ARFC B-Chain: DSWMEEVIKL CGRELVRAQI AICGMSTWS | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
6019 | |
Human Relaxin-2 | |
E. coli | |
RUO | |
bA12D24.1.1; bA12D24.1.2; H2; H2-preprorelaxin; H2-RLX; Prorelaxin H2; relaxin 2; Relaxin A chain; Relaxin B chain; relaxin H2; relaxin, ovarian, of pregnancy; RLN2; RLX H2-RLX; RLXH2 | |
Human | |
Recombinant | |
Lyophilized |