Learn More
ACROBiosystems Human Siglec-6 / CD327 Protein, Fc Tag
Marca: ACROBiosystems SI6-H5256-100UG
Detalles adicionales : Peso : 0.00001kg
Descripción
Sialic acid-binding Ig-like lectin 6 (SIGLEC6) is also known as CD antigen CD327, CD33 antigen-like 1 (CD33L or CD33L1), Obesity-binding protein 1 (OB-BP1). SIGLEC6 belongs to the immunoglobulin superfamily and SIGLEC (sialic acid binding Ig-like lectin) family, which contains two Ig-like C2-type (immunoglobulin-like) domains and one Ig-like V-type (immunoglobulin-like) domain. SIGLEC6 mediates sialic-acid dependent binding to cells. SIGLEC6 binds to alpha-2,6-linked sialic acid. The sialic acid recognition site may be masked by cis interactions with sialic acids on the same cell surface.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 60.3 kDa. As a result of glycosylation, the protein migrates as 80-100 kDa under reducing (R) condition, and 130-150 kDa and 250-300 kDa under non-reducing (NREspecificaciones
AAH35359 | |
100 μg | |
Gln 16 - Val 320 | |
SIGLEC6,CD327,CD33L,CD33L1,OBBP1,OB-BP1,CDw327 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
60.3 kDa | |
Human Siglec-6, Fc Tag (SI6-H5256) is expressed from human 293 cells (HEK293). It contains AA Gln 16 - Val 320 (Accession # AAH35359). | |
-20°C | |
Siglec-6 | |
Human | |
50 mM Tris, 100 mM Glycine, 150 mM NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |