Learn More
Invitrogen™ Human Troponin I ProQuantum Immunoassay Kit
The ProQuantum Human Troponin I Immunoassay Kit is designed to provide quantitative measurements of human Troponin I in small sample volumes.
Brand: Invitrogen™ A460745
Additional Details : Peso : 2.00000kg
Description
Utilizing proximity-based amplification technology, the assay combines the analyte specificity of high-affinity antibody-antigen binding with the signal detection and amplification capabilities of real-time PCR to achieve a simple yet powerful next-generation protein quantitation platform. A user-friendly workflow combined with intuitive software for analytics enables sample-to-answer in just 2 hours.
- High sensitivity-detect low levels of protein with greater sensitivity than traditional methods
- Broad dynamic range-≥5 logarithmic units, minimizing sample dilutions to ensure they fall within the range
- Small sample consumption-use 2–5 μL of sample (compared to 75 μL for triplicate wells with other methods)
- Fast, easy workflow-2 hours from sample to answer, with no wash steps
- No proprietary instrument to purchase-runs on any real-time PCR instrument
Specifications
troponina I cardíaca; isoforma de troponina I cardíaca; miocardiopatía dilatada 2 A (autosómica recesiva); CMD1FF; CMD2A; CMH7; cTNI; MGC116817; RCM1; Tn1; TnI; TNNC1; Tnni3; Tpn1; troponina 1, tipo 3; troponina I tipo 3 (cardíaca); troponina I, cardíaca 3; troponina I, músculo cardíaco; troponina I, similar a muscular cardíaco; troponina I3, tipo cardíaco | |
<0,3 pg/ml | |
Plasma, Sobrenadante, Suero | |
P19429 | |
Tampón de dilución de ensayo Conjugado de anticuerpos A Conjugado de anticuerpos B Tampón de dilución de conjugado de anticuerpos Estándar Mezcla maestra Ligasa | |
Para uso exclusivo en investigación | |
-20 °C | |
30 min |
De 0,32 a 25 000 pg/ml | |
ELISA | |
Instrumentos qPCR | |
TNNI3 | |
5 x 96 pruebas | |
Suero, 2-5 µl; plasma, 2-5 µl; sobrenadante, 2-5 µl | |
Troponina I cardíaca | |
2 h |