Learn More
R&D Systems™ Hyaluronan Quantikine ELISA Kit
Hyaluronan Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Marca: R&D Systems™ SHYAL0
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The Hyaluronan Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies multi-species Hyaluronan in cell culture supernates (50 ul), tissue homogenates (50 ul), serum (25 ul), edta plasma (25 ul), heparin plasma (25 ul).Especificaciones
0.63 to 40 ng/mL | |
HRP | |
35 kDa Hyaluronan in Human, Mouse, Rat, Canine, and Porcine Samples | |
1 Pack (6 Plates) | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
0.2 ng/mL | |
Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Tissue Homogenates | |
ELISA | |
Cell Culture Supernates (50 uL),Tissue Homogenates (50 uL),Serum (25 uL),EDTA Plasma (25 uL),Heparin Plasma (25 uL) |