Learn More
ACROBiosystems Influenza A [A/Shanghai/2/2013(H7N9)] Hemagglutinin (HA) Protein, Fc Tag
314.52€ - 2020.12€
Especificaciones
Número de acceso | AHF20558.1 |
---|---|
Peso molecular | 82.5 kDa |
Fuente | Influenza A [A/Shanghai/2/2013(H7N9)] HA, Fc Tag (HA9-V5253) is expressed from human 293 cells (HEK293). It contains AA Asp 19 - Gly 338, Leu 341 - Asp 523 (Accession # R4NN21-1). |
Inmunógeno | Asp 19 - Gly 338, Leu 341 - Asp 523 |
Requisitos de almacenamiento | -20°C |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15876917
|
ACROBiosystems
HA9-V5253-1MG |
1 mg |
2020.12€
1 miligramo |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
15866917
|
ACROBiosystems
HA9-V5253-100UG |
100 μg |
314.52€
100 microgramos |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
Influenza, commonly known as “the flu”, is an infectious disease of birds and mammals caused by RNA viruses of the family Orthomyxoviridae, the influenza viruses. The virus is divided into three main types (Influenzavirus A, Influenzavirus B, and Influenzavirus C), which are distinguished by differences in two major internal proteins (hemagglutinin (HA) and neuraminidase (NA), which are the most important targets for the immune system. The type A viruses are the most virulent human pathogens among the three influenza types and cause the most severe disease. The serotypes that have been confirmed in humans, ordered by the number of known human pandemic deaths, are: H1N1, H2N2, H3N2, H5N1, H7N7, H1N2, H9N2, H7N2, H7N3, H10N7, H7N9. H7N9 is a serotype of the species Influenzavirus A (avian influenza virus or bird flu virus). H7 normally circulates amongst avian populations with some variants known to occasionally infect humans. An H7N9 virus was first reported to have infected humans in 2013 in China.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 82.5 kDa. The protein migrates as 100-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
AHF20558.1 | |
Influenza A [A/Shanghai/2/2013(H7N9)] HA, Fc Tag (HA9-V5253) is expressed from human 293 cells (HEK293). It contains AA Asp 19 - Gly 338, Leu 341 - Asp 523 (Accession # R4NN21-1). | |
-20°C | |
Hemagglutinin (HA) | |
Influenza | |
50 mM Tris, 100 mM Glycine, pH7.5 | |
HEK293 | |
Viral Proteins |
82.5 kDa | |
Asp 19 - Gly 338, Leu 341 - Asp 523 | |
HA,Hemagglutinin | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |