Learn More
HMC Europe Cestas para autoclaves verticales
With this item you can easily and safely bring your sterilized goods into and out of the autoclave; we offer baskets in various sizes, with and without bowl or closed cans with and without holes.
Marca: HMC Europe 50027
Detalles adicionales : Peso : 0.50000kg
Descripción
For use with the HMC-HG and HMC-HV autoclaves.Especificaciones
11 in. | |
6.6 gal. | |
HV 25 autoclave | |
260 mm | |
Stainless Steel |
280 mm | |
25 L | |
10.23 in. | |
With collecting pan | |
280 x 260 mm |