missing translation for 'onlineSavingsMsg'
Learn More

L-Nipecotic acid, 96+%, Thermo Scientific Chemicals

Product Code. 15443998
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59045-82-8
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.159
MDL Number MFCD01630807
InChI Key XJLSEXAGTJCILF-YFKPBYRVSA-N
Synonym s-piperidine-3-carboxylic acid, s-+-nipecotic acid, s-nipecotic acid, 3s-piperidine-3-carboxylic acid, +-nipecotic acid, s-+-3-piperidinecarboxylic acid, 3s-nipecotic acid, l +-nipecotic acid, s-hexahydronicotinic acid, +-hexahydronicotinic acid
PubChem CID 6575389
ChEBI CHEBI:222169
IUPAC Name (3S)-piperidine-3-carboxylic acid
SMILES C1CC(CNC1)C(=O)O

Specifications

Melting Point 253°C to 257°C
Quantity 1 g
Formula Weight 129.16
Optical Rotation +5° (c=1 in Water)
Percent Purity ≥96%
Chemical Name or Material L-Nipecotic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'