Learn More
Kern Sohn Parabrisas de vidrio grande
Large glass draught shield with 3 sliding doors for easy access to the items being weighed.
Marca: Kern Sohn EG-A03
Detalles adicionales : Peso : 1.25000kg
Descripción
- Suitable for Precision Balances
- Balance Accessory
Especificaciones
Precision Balance with 118 mm Plate | |
Glass | |
140 x 150 x 130 mm |