Learn More
Lonza™ AccuGENE™ 1M Tris HCl Buffers
Ready-to-use solutions ideal for a wide range of molecular biology applications
Marca: Lonza™ 51236
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- Prepared with 18 megohm water
- Filtered using 0.2μ filter and filled into sterile bottles
- Manufactured according to strict quality control standards to ensure consistency
- Guaranteed DNase, RNase and protease-free
- Customized solutions are available to meet individual needs
Especificaciones
1 M | |
Ideal for a wide range of molecular biology applications | |
1000 mL |
Yes | |
7.2 |
Título del producto
For research or manufacturing use only.
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido