Learn More
Lovibond™ Celdas de colorimetría de vidrio de borosilicato
Celdas fusionadas de precisión
Marca: Lovibond™ 656030
Detalles adicionales : Peso : 0.06930kg
Descripción
Available in a range of dimensions and path lengths, made from optical glass to the highest standards. Note: For instruments equipped with a heater unit and whenever cells are subjected to thermal shock, it is recommended that borosilicate cells be used.
Series: W600 Borosilicate
Especificaciones
Colorimetry Cell | |
Model Fs, PFXi-880, 950 and 995 instruments with heated Samples (up to 100°C) | |
Borosilicate Glass | |
100 mm |