Learn More
Loxiglumide
CCK1 Receptor Antagonists
Marca: Tocris 3036/25
Detalles adicionales : Peso : 0.00970kg
Descripción
Loxiglumide is a CCK1 antagonist. Suppresses CCK-induced proliferation of acinar cells. Inhibits pancreatic secretion of digestive enzymes. Active in vivo.Especificaciones
107097-80-3 | |
25 mg | |
461.38 |
4-[(3,4-Dichlorobenzoyl)amino]-5-[(3-methoxypropyl)pentylamino]-5-oxopentanoic acid | |
C21H30Cl2N2O5 | |
97% |