Learn More
Eurofins DiscoverX Lyn Protein, Active
Active, mouse. Recombinant full-length mouse Lyn, containing an N-terminal His6-tag, for use in Kinase Assays.
Marca:  Eurofins DiscoverX 14-315
Encompass_Preferred
 Ver más versiones de este producto
Código de producto. 17360124
  • 525.15€ / 10 microgramos
Explore más ofertas especiales
Descripción
Descripción
The Lyn protein is a member of the Src family of non-receptor-associated protein-tyrosine kinases. LYN A differs from LYN B in having an additional 21 amino acids at the amino terminus. Lyn undergoes a concentration-dependent autophosphorylation, thus activating the catalytic site and decreasing the accessibility of the SH2 domain. Lyn phosphorylates and activates Tec, non-receptor tyrosine kinase that is coexpressed with Lyn and acts downstream of Lyn in intracellular signaling pathways. Lyn tyrosine kinase has been shown to be necessary for IgE-receptor (FceRI)-mediated mast-cell activation.
EMBL M64608, N425D as reported in SWISS PROT P25911
Especificaciones
Especificaciones
mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 0.1 mM EGTA, 150 mM NaCl, 0.03% Brij-35, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol, 5% glycerol. Frozen solution.
10 μg
Recombinant
Recombinant enzyme expressed in Sf21 insect cells
≥66%
60 kDa
Mouse
N-terminal 6His
Full-length
Lyn (mouse), active