Learn More
Eurofins DiscoverX Lyn Protein, Active
Active, mouse. Recombinant full-length mouse Lyn, containing an N-terminal His6-tag, for use in Kinase Assays.
525.15€ - 4008.15€
Especificaciones
Formulación | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 0.1 mM EGTA, 150 mM NaCl, 0.03% Brij-35, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol, 5% glycerol. Frozen solution. |
---|---|
Peso molecular | 60 kDa |
Especie | Mouse |
Recombinante | Recombinant |
Etiqueta de proteína | N-terminal 6His |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
17360124
|
Eurofins DiscoverX
14-315 |
10 μg |
525.15€
10 microgramos |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
17370124
|
Eurofins DiscoverX
14-315M |
250 μg |
4008.15€
250 microgramos |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Descripción
The Lyn protein is a member of the Src family of non-receptor-associated protein-tyrosine kinases. LYN A differs from LYN B in having an additional 21 amino acids at the amino terminus. Lyn undergoes a concentration-dependent autophosphorylation, thus activating the catalytic site and decreasing the accessibility of the SH2 domain. Lyn phosphorylates and activates Tec, non-receptor tyrosine kinase that is coexpressed with Lyn and acts downstream of Lyn in intracellular signaling pathways. Lyn tyrosine kinase has been shown to be necessary for IgE-receptor (FceRI)-mediated mast-cell activation.
EMBL M64608, N425D as reported in SWISS PROT P25911
Especificaciones
mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 0.1 mM EGTA, 150 mM NaCl, 0.03% Brij-35, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol, 5% glycerol. Frozen solution. | |
Mouse | |
N-terminal 6His | |
Full-length | |
Lyn (mouse), active |
60 kDa | |
Recombinant | |
Recombinant enzyme expressed in Sf21 insect cells | |
≥66% |