Learn More
Magnesium titanium oxide, 99% (metals basis)
Description
Magnesium titanium oxide is used in the primary chemical intermediates and in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 1610°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 1 kg |
| Solubility Information | Insoluble in water. |
| Formula Weight | 120.21 |
| Percent Purity | 99% |
| Odor | Odorless |
| Density | 3.36 g/mL |
| Assay | (metals basis) |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.