Learn More
Ácido metafosfórico, estabilizado, Thermo Scientific Chemicals
Ácido metafosfórico, 40 a 46 % (HPO3), HO3P, número de CAS-37267-86-0, 50813-16-6, hidrogenfosfonato, ácido metafosfórico, ácido meta-fosfórico, fosfonato de hidrógeno, ácido fosfénico, ácido m-fosfórico, ácido metafosfórico hpo3, fosfonato 1-, grupo x-fosfato, unii-mtk99r3uv0, 500 g, 5 ppm máx.
67.13€ - 220.10€
Identificadores de productos químicos
CAS | 37267-86-0, 50813-16-6 |
---|---|
Fórmula molecular | HO3P |
Peso molecular (g/mol) | 79.979 |
Número MDL | MFCD00011351 |
Clave InChI | UEZVMMHDMIWARA-UHFFFAOYSA-N |
Sinónimo | metaphosphoric acid, meta-phosphoric acid, hydrogenphosphonate, m-phosphoric acid, phosphenic acid, unii-mtk99r3uv0, metaphosphoric acid hpo3, x-phosphate-group, phosphonate 1-, hydrogen phosphonate |
PubChem CID | 3084658 |
SMILES | OP(=O)=O |
Código de producto | Marca | Cantidad | Envase | Precio | Cantidad y disponibilidad | ||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Envase | Precio | Cantidad y disponibilidad | ||||
10105040
|
Thermo Scientific Acros
219221000 |
100 g | Frasco de vidrio |
67.13€
100 g |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
10115040
|
Thermo Scientific Acros
219225000 |
500g | Frasco de vidrio |
220.10€
500 g |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identificadores de productos químicos
37267-86-0, 50813-16-6 | |
79.979 | |
UEZVMMHDMIWARA-UHFFFAOYSA-N | |
3084658 |
HO3P | |
MFCD00011351 | |
metaphosphoric acid, meta-phosphoric acid, hydrogenphosphonate, m-phosphoric acid, phosphenic acid, unii-mtk99r3uv0, metaphosphoric acid hpo3, x-phosphate-group, phosphonate 1-, hydrogen phosphonate | |
OP(=O)=O |
Especificaciones
Metaphosphoric acid | |
50 ppm máx. | |
(HPO3)n | |
MFCD00011351 | |
15, 7457 | |
Solubilidad en agua: soluble.Otras solubilidades: formación lenta del ortoácido, soluble en alcohol | |
OP(=O)=O | |
3084658 | |
40 to 46% (HPO3) |
37267-86-0, 50813-16-6 | |
HO3P | |
Masa vidriosa | |
01,665; 16,177 | |
metaphosphoric acid, meta-phosphoric acid, hydrogenphosphonate, m-phosphoric acid, phosphenic acid, unii-mtk99r3uv0, metaphosphoric acid hpo3, x-phosphate-group, phosphonate 1-, hydrogen phosphonate | |
UEZVMMHDMIWARA-UHFFFAOYSA-N | |
79.979 | |
79.98 |
Seguridad y manipulación
Declaración de GHS H:
Provoca graves quemaduras en la piel y lesiones oculares.
Puede irritar las vías respiratorias.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Peligro
missing translation for 'einecsNumber' : 253-433-4
missing translation for 'tsca' : TSCA
- Metaphosphoric acid
Palabra de advertencia
- Peligro
Clasificación
- Lesión ocular grave/irritación ocular Categoría 1
- Corrosión o irritación cutáneas Categoría 1B
- Toxicidad específica para determinados órganos Categoría 3
Indicaciones de peligro
- H318-Provoca lesiones oculares graves.
- H335-Puede irritar las vías respiratorias.
- H314-Provoca quemaduras graves en la piel y lesiones oculares graves.
Consejos de prudencia
- P280-Llevar guantes/prendas/gafas/máscara de protección.
- P301+P330+P331-EN CASO DE INGESTIÓN: Enjuagarse la boca. NO provocar el vómito.
- P303+P361+P353-EN CASO DE CONTACTO CON LA PIEL (o el pelo): Quitar inmediatamente todas las prendas contaminadas. Aclararse la piel con agua/ducharse.
- P304+P340-EN CASO DE INHALACIÓN: Transportar a la persona al aire libre y mantenerla en una posición que le facilite la respiración.
- P305+P351+P338-EN CASO DE CONTACTO CON LOS OJOS: Aclarar cuidadosamente con agua durante varios minutos. Quitar las lentes de contacto, si lleva y resulta fácil. Seguir aclarando.
- P310-Llamar inmediatamente a un CENTRO DE TOXICOLOG A/médico/.
RUO – Research Use Only