missing translation for 'onlineSavingsMsg'
Learn More

Propionato de metilo, 99+ %, Thermo Scientific Chemicals

41.51€ - 122.61€

Identificadores de productos químicos

CAS 554-12-1
Fórmula molecular C4H8O2
Peso molecular (g/mol) 88.11
Número MDL MFCD00009306
Clave InChI RJUFJBKOKNCXHH-UHFFFAOYSA-N
Sinónimo methyl propionate, propanoic acid, methyl ester, methyl propylate, methylpropionate, propionic acid, methyl ester, propionate de methyle, propanoic acid methyl ester, fema number 2742, propionic acid methyl ester, methyl propionate natural
PubChem CID 11124
Nombre IUPAC propanoato de metilo
SMILES CCC(=O)OC
Ver más especificaciones

Products 2
Product Code Brand Cantidad Envase Price Quantity & Availability  
Product Code Brand Cantidad Envase Price Quantity & Availability  
10295430
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Acros
149052500
250 mL Frasco de vidrio
41.51€
250 ml
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
10021570
missing translation for 'viewDocuments'
Thermo Scientific Acros
149050010
1 L Frasco de vidrio
122.61€
1 litro
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

554-12-1
88.11
RJUFJBKOKNCXHH-UHFFFAOYSA-N
11124
CCC(=O)OC
C4H8O2
MFCD00009306
methyl propionate, propanoic acid, methyl ester, methyl propylate, methylpropionate, propionic acid, methyl ester, propionate de methyle, propanoic acid methyl ester, fema number 2742, propionic acid methyl ester, methyl propionate natural
propanoato de metilo

Specifications

Methyl propionate
1mg KOH/g max.
-88°C
0.9150g/mL
-2°C
99% min. (GC)
C2H5CO2CH3
MFCD00009306
15, 6185
methyl propionate, propanoic acid, methyl ester, methyl propylate, methylpropionate, propionic acid, methyl ester, propionate de methyle, propanoic acid methyl ester, fema number 2742, propionic acid methyl ester, methyl propionate natural
RJUFJBKOKNCXHH-UHFFFAOYSA-N
propanoato de metilo
11124
99+%
99+%
554-12-1
Incolora
78°C to 79°C
Auténtico
C4H8O2
1.377
02, 239
0.915
Solubility in water: 5g/L water (20°C). Other solubilities: miscible with alcohol and ether,soluble in most common organic solvents
CCC(=O)OC
88.11
88.11
Líquido
Videos
Safety and Handling

Safety and Handling

Declaración de GHS H:
Nocivo en caso de inhalación.
Líquido y vapores muy inflamables.

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Palabra de aviso de GHS:Peligro

missing translation for 'einecsNumber' : 209-060-4

missing translation for 'rtecsNumber' : UF5970000.

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Certificates

RUO – Research Use Only