Learn More
Kern Sohn Cámara de microscopio ODC
Specialists in microscopy for measurement, counting, documentation, archiving and image processing.
Marca: Kern Sohn ODC 841
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- The universal microscope cameras can be used anywhere and can be connected to the microscope as well as to a laptop or PC using the USB cable (USB 2.0)
- Through the proven CMOS technology, in connection with the USB 2.0 the images are shown quickly and clearly
- These cameras are also ideal for more demanding applications, such as, for example, darkfield, phase contrast and for fluorescence applications.
- Your daily work is made significantly easier with the very best synchronisation, a high frame rate as well as stable image performance together with our Microscope VIS software which we deliver with the product.
- The power supply is through the USB cable, which means that no additional power supply is required
- As well as the camera, the delivery includes our multi-lingual KERN Microscope VIS Basic KERN OXM 901, a USB cable (Length: 2000 mm) various eyepiece adapters and an object micrometer to calibrate the software.
- Please order the appropriate C-mount adapter to fit your KERN microscope now.
Especificaciones
Microscopios | |
20 MP | |
1 in. Sensor |
USB 2.0 | |
CMOS |