Learn More
AGER, Mouse, Clone: 1C5, Abnova™
Mouse monoclonal antibody raised against recombinant AGER.
Marca: Abnova MAB1724.50ug
Descripción
This gene encodes a member of the immunoglobulin superfamily of cell surface molecules. It is a receptor for various molecules, including the amyloidogenic form of serum amyloid A, amyloid-beta protein, members of the S100/calgranulin superfamily and advanced glycation end products. The gene lies within the major histocompatibility complex (MHC) class III region on chromosome 6. Alternative splicing results in two transcript variants encoding different isoforms. [provided by RefSeq
Especificaciones
AGER | |
Monoclonal | |
Unconjugated | |
Western Blot (0.1 ug/mL) Immunocytochemistry (1 ug/mL) ELISA (0.1 ug/mL) Flow Cyt (1 ug/mL) The optimal working dilution should be determined by the end user. | |
AGER | |
AGER | |
Recombinant protein corresponding to human AGER. | |
RUO | |
177 | |
Liquid |
ELISA, Flow Cytometry, Immunocytochemistry, Western Blot | |
1C5 | |
Mouse monoclonal antibody raised against recombinant AGER. | |
In PBS (0.1% proclin, 2.0% Block Ace) | |
MGC22357/RAGE | |
Mouse | |
50 μg | |
Primary | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 κ |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.